Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 2-chloro-3-fluoro-4-pyridinecarboxylate ≥97%

Supplier: AOB CHEM USA
Warning

34853-1G 34853-25G 34853-500MG 34853-5G
76941-004EA 379.81 USD InStock
76941-004 76941-006 76941-008 76941-010
Ethyl 2-chloro-3-fluoro-4-pyridinecarboxylate ≥97%
Ethyl 2-chloro-3-fluoro-4-pyridinecarboxylate
Precaución: for Research Use Only


Formula: C₈H₇ClFNO₂
Storage Temperature: Ambient
MDL Number: MFCD14698350
CAS Number: 1214351-53-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty