Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 6-bromo-3-chloro-2-fluorobenzoate ≥97%

Supplier: AOB CHEM USA
Warning

35210-1G 35210-25G 35210-500MG 35210-5G
76941-148EA 260.99 USD InStock
76941-148 76941-150 76941-152 76941-154
Methyl 6-bromo-3-chloro-2-fluorobenzoate ≥97%
Methyl 6-bromo-3-chloro-2-fluorobenzoate
Precaución: for Research Use Only


Formula: C₈H₅BrClFO₂
Storage Temperature: Refrigerator
MDL Number: MFCD27978532
CAS Number: 1505850-20-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty