Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 5-fluoro-2-(trifluoromethyl)benzoate ≥95%

Supplier: AOB CHEM USA
Warning

35464-1G 35464-25G 35464-500MG 35464-5G
76941-208EA 331.01 USD InStock
76941-208 76941-210 76941-212 76941-214
Methyl 5-fluoro-2-(trifluoromethyl)benzoate ≥95%
Methyl 5-fluoro-2-(trifluoromethyl)benzoate
Precaución: for Research Use Only


Formula: C₉H₆F₄O₂
MW: 222.136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06203755
CAS Number: 773873-90-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty