Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4-bromo-2-isopropylbenzoate ≥97%

Supplier: AOB CHEM USA
Warning

35529-1G 35529-25G 35529-500MG 35529-5G
76941-224EA 296 USD InStock
76941-224 76941-226 76941-228 76941-230
Methyl 4-bromo-2-isopropylbenzoate ≥97%
Methyl 4-bromo-2-isopropylbenzoate
Precaución: for Research Use Only


Formula: C₁₁H₁₃BrO₂
MW: 257.124 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD30295010
CAS Number: 643094-19-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty