Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 2,6-Difluoro-5-formylbenzoate ≥97%

Supplier: AOB CHEM USA
Warning

35542-1G 35542-25G 35542-500MG 35542-5G
76941-240EA 256.74 USD InStock
76941-240 76941-242 76941-244 76941-246
Ethyl 2,6-Difluoro-5-formylbenzoate ≥97%
Ethyl 2,6-Difluoro-5-formylbenzoate
Precaución: for Research Use Only


Formula: C₁₀H₈F₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD32631810
CAS Number: 2271442-96-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty