Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 3-bromo-4-iodo-5-methylbenzoate ≥97%

Supplier: AOB CHEM USA
Warning

35857-1G 35857-25G 35857-500MG 35857-5G
76941-318EA 260.99 USD InStock
76941-318 76941-320 76941-322 76941-324
Methyl 3-bromo-4-iodo-5-methylbenzoate ≥97%
Methyl 3-bromo-4-iodo-5-methylbenzoate
Precaución: for Research Use Only


Formula: C₉H₈BrIO₂
MW: 354.967 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11846083
CAS Number: 1160574-57-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty