Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Ethoxy-2-naphthoic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

36898-1G 36898-25G 36898-500MG 36898-5G
76941-620EA 195.21 USD InStock
76941-620 76941-622 76941-624 76941-626
6-Ethoxy-2-naphthoic acid ≥97% research grade
6-Ethoxy-2-naphthoic acid


Formula: C₁₃H₁₂O₃
MW: 216.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18815314
CAS Number: 614754-39-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty