Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Isopropyl-2-methoxybenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

37064-1G 37064-25G 37064-500MG 37064-5G
76941-662EA 370.26 USD InStock
76941-662 76941-664 76941-666 76941-668
5-Isopropyl-2-methoxybenzoic acid ≥97%
5-Isopropyl-2-methoxybenzoic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₄O₃
Storage Temperature: Refrigerator
MDL Number: MFCD01242354
CAS Number: 68014-67-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty