Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 2-bromo-5-fluorobenzoate ≥97% research grade

Supplier: AOB CHEM USA
Warning

37428-1G 37428-25G 37428-500MG 37428-5G
76941-800EA 124.13 USD InStock
76941-800 76941-802 76941-804 76941-806
tert-Butyl 2-bromo-5-fluorobenzoate ≥97% research grade
tert-Butyl 2-bromo-5-fluorobenzoate


Formula: C₁₁H₁₂BrFO₂
MW: 275.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17676529
CAS Number: 1263281-14-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty