Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-2-chloro-3-methoxybenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

37563-1G 37563-25G 37563-500MG 37563-5G
76941-850EA 225.98 USD InStock
76941-850 76941-852 76941-854 76941-856
6-Bromo-2-chloro-3-methoxybenzoic acid ≥97%
6-Bromo-2-chloro-3-methoxybenzoic acid


Formula: C₈H₆BrClO₃
Storage Temperature: Refrigerator
MDL Number: MFCD30829706
CAS Number: 1055968-54-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty