Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 6-bromo-2-chloro-3-methoxybenzoate ≥97% research grade

Supplier: AOB CHEM USA
Warning

38094-1G 38094-25G 38094-500MG 38094-5G
76942-076EA 379.81 USD InStock
76942-076 76942-078 76942-080 76942-082
Methyl 6-bromo-2-chloro-3-methoxybenzoate ≥97% research grade
Methyl 6-bromo-2-chloro-3-methoxybenzoate


Formula: C₉H₈BrClO₃
MW: 279.515 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22689511
CAS Number: 750586-08-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty