Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 3-fluoro-4-isopropoxybenzoate ≥97% research grade

Supplier: AOB CHEM USA
Warning

38619-1G 38619-25G 38619-500MG 38619-5G
76942-276EA 144.29 USD InStock
76942-276 76942-278 76942-280 76942-282
Ethyl 3-fluoro-4-isopropoxybenzoate ≥97% research grade
Ethyl 3-fluoro-4-isopropoxybenzoate


Formula: C₁₂H₁₅FO₃
Storage Temperature: Ambient
MDL Number: MFCD32707101
CAS Number: 2432849-03-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty