Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-2-fluoro-3-isopropoxybenzoic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

38625-1G 38625-25G 38625-500MG 38625-5G
76942-290EA 183.54 USD InStock
76942-290 76942-292 76942-294 76942-296
6-Bromo-2-fluoro-3-isopropoxybenzoic acid ≥97% research grade
6-Bromo-2-fluoro-3-isopropoxybenzoic acid


Formula: C₁₀H₁₀BrFO₃
MW: 277.087 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32707103
CAS Number: 2413441-26-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty