Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 6-fluoro-2,3-dimethoxybenzoate ≥97%

Supplier: AOB CHEM USA
Warning

45900-1G 45900-500MG 45900-5G
76942-502EA 558.04 USD InStock
76942-502 76942-504 76942-506
Ethyl 6-fluoro-2,3-dimethoxybenzoate ≥97%
Ethyl 6-fluoro-2,3-dimethoxybenzoate


Formula: C₁₁H₁₃FO₄
Storage Temperature: Ambient
MDL Number: MFCD16038838
CAS Number: 1260651-11-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty