Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-(Benzyloxy)-2-methylnicotinic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

46086-1G 46086-25G 46086-500MG 46086-5G
76942-568EA 206.88 USD InStock
76942-568 76942-570 76942-572 76942-574
6-(Benzyloxy)-2-methylnicotinic acid ≥97% research grade
6-(Benzyloxy)-2-methylnicotinic acid


Formula: C₁₄H₁₃NO₃
MW: 243.258 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21089095
CAS Number: 1355172-95-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty