Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-hydroxy-5-(trifluoromethoxy)benzoate ≥97% research grade

Supplier: AOB CHEM USA
Warning

47430-1G 47430-25G 47430-500MG 47430-5G
76942-692EA 147.47 USD InStock
76942-692 76942-694 76942-696 76942-698
Methyl 2-hydroxy-5-(trifluoromethoxy)benzoate ≥97% research grade
Methyl 2-hydroxy-5-(trifluoromethoxy)benzoate


Formula: C₉H₇F₃O₄
Storage Temperature: Refrigerator
MDL Number: MFCD00219949
CAS Number: 175204-89-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty