Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4-bromo-2,3-dichlorobenzoate ≥97% research grade

Supplier: AOB CHEM USA
Warning

48059-1G 48059-25G 48059-5G
76942-728EA 109.28 USD InStock
76942-728 76942-730 76942-732
Methyl 4-bromo-2,3-dichlorobenzoate ≥97% research grade
Methyl 4-bromo-2,3-dichlorobenzoate


Formula: C₈H₅BrCl₂O₂
MW: 283.934 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD30530457
CAS Number: 2055839-96-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty