Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4-isopropoxy-3-methoxybenzoate ≥97%

Supplier: AOB CHEM USA
Warning

48619-1G 48619-25G 48619-500MG 48619-5G
76942-772EA 215.37 USD InStock
76942-772 76942-774 76942-776 76942-778
Methyl 4-isopropoxy-3-methoxybenzoate ≥97%
Methyl 4-isopropoxy-3-methoxybenzoate


Formula: C₁₂H₁₆O₄
Storage Temperature: Ambient
MDL Number: MFCD5663675
CAS Number: 3535-27-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty