Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 5-bromo-2,4-dichlorobenzoate ≥97%

Supplier: AOB CHEM USA
Warning

49262-1G 49262-25G 49262-500MG 49262-5G
76942-868EA 206.88 USD InStock
76942-868 76942-870 76942-872 76942-874
Methyl 5-bromo-2,4-dichlorobenzoate ≥97%
Methyl 5-bromo-2,4-dichlorobenzoate
Precaución: for Research Use Only


Formula: C₈H₅BrCl₂O₂
MW: 283.934 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12032203
CAS Number: 1305712-91-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty