Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(2-Bromo-5-fluorophenyl)-2,2,2-trifluoroacetamide ≥95%

Supplier: AOB CHEM USA
Warning

51854-1G 51854-25G 51854-500MG 51854-5G
76943-006EA 128.37 USD InStock
76943-006 76943-008 76943-010 76943-012
N-(2-Bromo-5-fluorophenyl)-2,2,2-trifluoroacetamide ≥95%
N-(2-Bromo-5-fluorophenyl)-2,2,2-trifluoroacetamide


Formula: C₈H₄BrF₄NO
Storage Temperature: Ambient
MDL Number: MFCD30723605
CAS Number: 1882371-64-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty