Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-2-fluoro-3-formylbenzoic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

52071-1G 52071-25G 52071-500MG 52071-5G
76943-022EA 173.99 USD InStock
76943-022 76943-024 76943-026 76943-028
6-Bromo-2-fluoro-3-formylbenzoic acid ≥97% research grade
6-Bromo-2-fluoro-3-formylbenzoic acid


Formula: C₈H₄BrFO₃
MW: 247.018 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD34564212
CAS Number: 2092783-38-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty