Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 3-bromobenzo[b]thiophene-2-carboxylate ≥97%

Supplier: AOB CHEM USA
Warning

54345-1G 54345-25G 54345-500MG 54345-5G
76943-140EA 287.51 USD InStock
76943-140 76943-142 76943-144 76943-146
Methyl 3-bromobenzo[b]thiophene-2-carboxylate ≥97%
Methyl 3-bromobenzo[b]thiophene-2-carboxylate


Formula: C₁₀H₇BrO₂S
Storage Temperature: Refrigerator
MDL Number: MFCD20526506
CAS Number: 34128-30-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty