Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 3-hydroxyheptanoate ≥95%

Supplier: AOB CHEM USA
Warning

55613-1G 55613-25G 55613-500MG 55613-5G
76943-248EA 233.4 USD InStock
76943-248 76943-250 76943-252 76943-254
Methyl 3-hydroxyheptanoate ≥95%
Methyl 3-hydroxyheptanoate


Formula: C₈H₁₆O₃
Storage Temperature: Refrigerator
MDL Number: MFCD25959586
CAS Number: 15889-95-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty