Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4-bromo-2-fluoro-3-methoxybenzoate ≥97% research grade

Supplier: AOB CHEM USA
Warning

55879-1G 55879-25G 55879-500MG 55879-5G
76943-286EA 302.36 USD InStock
76943-286 76943-288 76943-290 76943-292
Methyl 4-bromo-2-fluoro-3-methoxybenzoate ≥97% research grade
Methyl 4-bromo-2-fluoro-3-methoxybenzoate


Formula: C₉H₈BrFO₃
MW: 263.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23707520
CAS Number: 908248-31-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty