Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 5-bromo-2-cyanobenzoate ≥95% research grade

Supplier: AOB CHEM USA
Warning

57349-1G 57349-25G 57349-500MG 57349-5G
76943-382EA 393.6 USD InStock
76943-382 76943-384 76943-386 76943-388
Ethyl 5-bromo-2-cyanobenzoate ≥95% research grade
Ethyl 5-bromo-2-cyanobenzoate


Formula: C₁₀H₈BrNO₂
MW: 254.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11041400
CAS Number: 127510-93-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty