Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-(4-Fluorophenyl)nicotinic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

58048-1G 58048-25G 58048-5G
76943-456EA 115.64 USD InStock
76943-456 76943-458 76943-460
6-(4-Fluorophenyl)nicotinic acid ≥97% research grade
6-(4-Fluorophenyl)nicotinic acid


Formula: C₁₂H₈FNO₂
MW: 217.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06410388
CAS Number: 223127-24-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty