Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-bromo-4-fluoro-5-methoxybenzoate ≥97%

Supplier: AOB CHEM USA
Warning

59891-1G 59891-25G 59891-500MG 59891-5G
76943-546EA 124.13 USD InStock
76943-546 76943-548 76943-550 76943-552
Methyl-2-bromo-4-fluoro-5-methoxybenzoate ≥97%
Methyl-2-bromo-4-fluoro-5-methoxybenzoate


Formula: C₉H₈BrFO₃
Storage Temperature: Ambient
MDL Number: MFCD20482372
CAS Number: 1007455-22-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty