Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 3-bromo-5-isopropyl-2-(methylthio)benzoate ≥95% research grade

Supplier: AOB CHEM USA
Warning

61410-1G 61410-500MG 61410-5G
76943-630EA 560.16 USD InStock
76943-630 76943-632 76943-634
Methyl 3-bromo-5-isopropyl-2-(methylthio)benzoate ≥95% research grade
Methyl 3-bromo-5-isopropyl-2-(methylthio)benzoate


Formula: C₁₂H₁₅BrO₂S
MW: 303.2153 g/mol
Storage Temperature: Ambient
CAS Number: 2918886-84-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty