Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 4-methoxy-2,6-dimethylbenzoate ≥97% research grade

Supplier: AOB CHEM USA
Warning

62099-1G 62099-25G 62099-500MG 62099-5G
76943-658EA 272.66 USD InStock
76943-658 76943-660 76943-662 76943-664
Ethyl 4-methoxy-2,6-dimethylbenzoate ≥97% research grade
Ethyl 4-methoxy-2,6-dimethylbenzoate


Formula: C₁₂H₁₆O₃
MW: 208.2536 g/mol
Storage Temperature: Ambient
CAS Number: 412949-76-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty