Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(4-Bromo-2,5-difluorophenyl)acetamide ≥97%

Supplier: AOB CHEM USA
Warning

65250-1G 65250-25G 65250-5G
76943-784EA 79.57 USD InStock
76943-784 76943-786 76943-788
N-(4-Bromo-2,5-difluorophenyl)acetamide ≥97%
N-(4-Bromo-2,5-difluorophenyl)acetamide


Formula: C₈H₆BrF₂NO
Storage Temperature: Ambient
CAS Number: 1065100-92-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty