Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 4'-formyl-[1,1'-biphenyl]-4-carboxylate ≥97%

Supplier: AOB CHEM USA
Warning

72518-1G 72518-25G 72518-500MG 72518-5G
76943-824EA 342.68 USD InStock
76943-824 76943-826 76943-828 76943-830
Ethyl 4'-formyl-[1,1'-biphenyl]-4-carboxylate ≥97%
Ethyl 4'-formyl-[1,1'-biphenyl]-4-carboxylate


Formula: C₁₆H₁₄O₃
Storage Temperature: Refrigerator
MDL Number: MFCD06738980
CAS Number: 70917-02-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty