Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 2-[3-(trifluoromethyl)phenyl]glyoxylate ≥97% research grade

Supplier: AOB CHEM USA
Warning

82397-1G 82397-25G 82397-5G
76943-950EA 116.7 USD InStock
76943-950 76943-952 76943-954
Ethyl 2-[3-(trifluoromethyl)phenyl]glyoxylate ≥97% research grade
Ethyl 2-[3-(trifluoromethyl)phenyl]glyoxylate


Formula: C₁₁H₉F₃O₃
Storage Temperature: Refrigerator
MDL Number: MFCD01934868
CAS Number: 110193-60-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty