Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-3,4-methylenedioxybenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

87623-1G 87623-25G 87623-500MG 87623-5G
76944-100EA 144.29 USD InStock
76944-100 76944-102 76944-104 76944-106
6-Bromo-3,4-methylenedioxybenzoic acid ≥97%
6-Bromo-3,4-methylenedioxybenzoic acid
Precaución: for Research Use Only


Formula: C₈H₅BrO₄
Storage Temperature: Ambient
MDL Number: MFCD00156994
CAS Number: 60546-62-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty