Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-2-chloro-3-(methylthio)pyridine ≥97% research grade

Supplier: AOB CHEM USA
Warning

91265-1G 91265-25G 91265-500MG 91265-5G
76944-136EA 223.85 USD InStock
76944-136 76944-138 76944-140 76944-142
6-Bromo-2-chloro-3-(methylthio)pyridine ≥97% research grade
6-Bromo-2-chloro-3-(methylthio)pyridine


Formula: C₆H₅BrClNS
MW: 238.533 g/mol
Storage Temperature: Ambient
CAS Number: 2921837-38-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty