Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 3-hydroxydecanoate ≥95% research grade

Supplier: AOB CHEM USA
Warning

55616-1G 55616-25G 55616-500MG 55616-5G
76943-256EA 201.58 USD InStock
76943-256 76943-258 76943-260 76943-262
Methyl 3-hydroxydecanoate ≥95% research grade
Methyl 3-hydroxydecanoate


Formula: C₁₁H₂₂O₃
MW: 202.291 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD133276
CAS Number: 62675-82-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty