Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Methyl-2-iodo-4-hydroxybenzene ≥97%

Supplier: AOB CHEM USA
Warning

21757-1G 21757-25G 21757-500MG 21757-5G
76884-846EA 137.92 USD InStock
76884-846 76884-848 76884-850 76884-852
1-Methyl-2-iodo-4-hydroxybenzene ≥97%
1-Methyl-2-iodo-4-hydroxybenzene


Formula: C₇H₇IO
Storage Temperature: Refrigerator
MDL Number: MFCD09261218
CAS Number: 626250-54-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty