Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-2,6-dibromoanisole ≥97%

Supplier: AOB CHEM USA
Warning

38602-1G 38602-25G 38602-500MG 38602-5G
76889-662EA 213.25 USD InStock
76889-662 76889-664 76889-666 76889-668
4-Chloro-2,6-dibromoanisole ≥97%
4-Chloro-2,6-dibromoanisole
Precaución: for Research Use Only


Formula: C₇H₅Br₂ClO
MW: 300.3750 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00068569
CAS Number: 174913-44-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty