Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dibromo-6-chloro-3-fluoroaniline ≥97%

Supplier: AOB CHEM USA
Warning

62257-1G 62257-25G 62257-500MG 62257-5G
76892-566EA 213.25 USD InStock
76892-566 76892-568 76892-570 76892-572
2,4-Dibromo-6-chloro-3-fluoroaniline ≥97%
2,4-Dibromo-6-chloro-3-fluoroaniline


Formula: C₆H₃Br₂ClFN
Storage Temperature: Ambient
CAS Number: 1823495-88-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty