Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Borono-2,3-dichlorobenzoic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

45571-1G 45571-25G 45571-500MG 45571-5G
76916-366EA 291.75 USD InStock
76916-366 76916-368 76916-370 76916-372
4-Borono-2,3-dichlorobenzoic acid ≥97% research grade
4-Borono-2,3-dichlorobenzoic acid


Formula: C₇H₅BCl₂O₄
Storage Temperature: Refrigerator
MDL Number: MFCD32065679
CAS Number: 2351934-06-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty