Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4-fluorophenetole 95% research grade

Supplier: AOB CHEM USA
Warning

16787-1G 16787-25G 16787-5G 16787-500MG
76896-596EA 102.91 USD InStock
76896-596 76896-598 76896-600 77648-886
2-Chloro-4-fluorophenetole 95% research grade
2-Chloro-4-fluorophenetole


Formula: C₈H₈ClFO
MW: 174.6 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00672985
CAS Number: 181305-71-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty