Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxyphenyl acetate ≥97%

Supplier: AOB CHEM USA
Warning

16103-1G 16103-25G 16103-500MG 16103-5G
76925-640EA 162.32 USD InStock
76925-640 76925-642 76925-644 76925-646
4-Methoxyphenyl acetate ≥97%
4-Methoxyphenyl acetate


Formula: C₉H₁₀O₃
Storage Temperature: Refrigerator
MDL Number: MFCD00048211
CAS Number: 1200-06-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty