Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-(phenylmethyl)-2H-indazole ≥97%

Supplier: AOB CHEM USA
Warning

23088-1G 23088-25G 23088-500MG 23088-5G
76927-932EA 171.87 USD InStock
76927-932 76927-934 76927-936 76927-938
5-Bromo-2-(phenylmethyl)-2H-indazole ≥97%
5-Bromo-2-(phenylmethyl)-2H-indazole


Formula: C₁₄H₁₁BrN₂
Storage Temperature: Refrigerator
MDL Number: MFCD22056752
CAS Number: 937049-51-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty