Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-methyl-4-(trifluoromethyl)benzoate ≥97%

Supplier: AOB CHEM USA
Warning

29984-1G 29984-25G 29984-500MG 29984-5G
76939-540EA 290.69 USD InStock
76939-540 76939-542 76939-544 76939-546
Methyl 2-methyl-4-(trifluoromethyl)benzoate ≥97%
Methyl 2-methyl-4-(trifluoromethyl)benzoate


Formula: C₁₀H₉F₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD14698067
CAS Number: 341030-79-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty