Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 5-bromo-2-chloronicotinate ≥97% research grade

Supplier: AOB CHEM USA
Warning

30342-25G 30342-5G
76939-656EA 418 USD InStock
76939-656 76939-658
Ethyl 5-bromo-2-chloronicotinate ≥97% research grade
Ethyl 5-bromo-2-chloronicotinate


Formula: C₈H₇BrClNO₂
MW: 264.50 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14698313
CAS Number: 1214332-67-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty