Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-5-(1,3-dioxolan-2-yl)-2-methoxybenzene ≥97%

Supplier: AOB CHEM USA
Warning

11150-1G 11150-25G 11150-500MG 11150-5G
76882-136EA 171.87 USD InStock
76882-136 76882-138 76882-140 76882-142
1-Bromo-5-(1,3-dioxolan-2-yl)-2-methoxybenzene ≥97%
1-Bromo-5-(1,3-dioxolan-2-yl)-2-methoxybenzene


Formula: C₁₀H₁₁BrO₃
Storage Temperature: Refrigerator
MDL Number: MFCD01318959
CAS Number: 223418-72-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty