Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dibromo-5-methoxyphenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

11695-1G 11695-25G 11695-500MG 11695-5G
76882-258EA 135.8 USD InStock
76882-258 76882-260 76882-262 76882-264
2,4-Dibromo-5-methoxyphenylboronic acid ≥97%
2,4-Dibromo-5-methoxyphenylboronic acid
Precaución: for Research Use Only


Formula: C₇H₇BBr₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD06659860
CAS Number: 89677-46-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty