Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-Indazole-5-boronic acid pinacol ester ≥97%

Supplier: AOB CHEM USA
Warning

13212-1G 13212-25G 13212-5G
76882-588EA 66.84 USD InStock
76882-588 76882-590 76882-592
1H-Indazole-5-boronic acid pinacol ester ≥97%
1H-Indazole-5-boronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₃H₁₇BN₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD07367523
CAS Number: 862723-42-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty