Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Iodo-2-phenoxybenzene ≥97% research grade

Supplier: AOB CHEM USA
Warning

134666-1G 134666-25G 134666-5G
76882-616EA 63.66 USD InStock
76882-616 76882-618 76882-620
1-Iodo-2-phenoxybenzene ≥97% research grade
1-Iodo-2-phenoxybenzene


Formula: C₁₂H₉IO
MW: 296.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00767196
CAS Number: 34883-46-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty