Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2-Bromo-5-iodophenyl)-1,3-dioxolane ≥97%

Supplier: AOB CHEM USA
Warning

17065-1G 17065-25G 17065-500MG 17065-5G
76883-572EA 181.42 USD InStock
76883-572 76883-574 76883-576 76883-578
2-(2-Bromo-5-iodophenyl)-1,3-dioxolane ≥97%
2-(2-Bromo-5-iodophenyl)-1,3-dioxolane


Formula: C₉H₈BrIO₂
Storage Temperature: Refrigerator
MDL Number: MFCD31619383
CAS Number: 2221812-24-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty