Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5,6-Difluorosalicylaldehyde ≥97%

Supplier: AOB CHEM USA
Warning

17241-1G 17241-25G 17241-500MG 17241-5G
76883-650EA 147.47 USD InStock
76883-650 76883-652 76883-654 76883-656
5,6-Difluorosalicylaldehyde ≥97%
5,6-Difluorosalicylaldehyde
Precaución: for Research Use Only


Formula: C₇H₄F₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD08275282
CAS Number: 187543-89-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty